Få mere at vide
[1,4'-Bipiperidine]-1'-carbonyl-d10-1'-carboxylic Acid tert-Butyl Ester, TRC
Artikelnummer. 30335602
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30335602 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335602 Leverandør Toronto Research Chemicals Leverandørnr. TRCB398972100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale [1,4'-Bipiperidine]-1'-carbonyl-d10-1'-carboxylic Acid tert-Butyl Ester
CAS 718613-18-4
Molekylær formel C15H18D10N2O2
Mængde 100 mg
Synonym [1,4'-Bipiperidine-2,2,3,3,4,4,5,5,6,6-d10]-1'-carboxylic Acid 1,1-Dimethylethyl Ester
InChI formel InChI=1S/C15H28N2O2/c1-15(2,3)19-14(18)17-11-7-13(8-12-17)16-9-5-4-6-10-16/h13H,4-12H2,1-3H3/i4D2,5D2,6D2,9D2,10D2
SMIL [2H]C1([2H])N(C2CCN(CC2)C(=O)OC(C)(C)C)C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]
IUPAC navn tert-butyl 4-(2,2,3,3,4,4,5,5,6,6-decadeuteriopiperidin-1-yl)piperidine-1-carboxylate
Molekylvægt (g/mol) 278.46
Formel vægt 278.2778