missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Benzenedimethanol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.
Solubility
Soluble in ether, toluene, methanol and hot water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
1,4-Benzenedimethanol is used as an organic synthesis intermediate. It is also used for preparation of soluble polyphenylene and p-tolyl-methanol.
Solubility
Soluble in ether, toluene, methanol and hot water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Specifications
Melting Point | 117°C to 121°C |
Boiling Point | 138°C to 143°C (1 mmHg) |
Quantity | 100 g |
Beilstein | 2042077 |
Solubility Information | Soluble in ether,toluene,methanol and hot water. |
Formula Weight | 138.17 |
Percent Purity | 99% |
Chemical Name or Material | 1,4-Benzenedimethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'