Learn More
1,4-Benzenedicarboxylic Acid Dimethyl Ester, TRC
Product Code. 30343991
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity Unit Size
30343991 1 g 1g
1 options
Product Code. 30343991 Supplier Toronto Research Chemicals Supplier No. TRCB1853251G
 Restricted Item
Forventet forsendelse: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Terephthalic acid-bis-methyl ester
CAS 120-61-6
Molecular Formula C10 H10 O4
Quantity 1 g
Synonym 1,4-Benzenedicarboxylic acid, dimethyl ester (9CI), Terephthalic acid, dimethyl ester (6CI,7CI,8CI), DMT, Dimethyl 1,4-benzenedicarboxylate, Dimethyl p-benzenedicarboxylate, Dimethyl p-phthalate, Dimethyl terephthalate, Methyl 4-(carbomethoxy)benzoate, Methyl p-(methoxycarbonyl)benzoate, NSC 3503
InChI Formula InChI=1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3
SMILES COC(=O)c1ccc(cc1)C(=O)OC
IUPAC Name dimethyl benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 194.18
Formula Weight 194.0579