missing translation for 'onlineSavingsMsg'
Få mere at vide

1,4-Benzenedicarboxylic Acid-d4 Dimethyl Ester, TRC

Artikelnummer. 30340397
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30340397 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30340397 Leverandør Toronto Research Chemicals Leverandørnr. TRCB185327250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,4-Benzenedicarboxylic Acid-d4 Dimethyl Ester
CAS 74097-01-9
Molekylær formel C10 D4 H6 O4
Mængde 250 mg
Synonym Terephthalic Acid-d4 Dimethyl Ester, DMT-d4, Dimethyl 1,4-Benzenedicarboxylate-d4, Dimethyl p-Benzenedicarboxylate-d4, Dimethyl p-Phthalate-d4, Dimethyl Terephthalate-d4, Methyl 4-(Carbomethoxy)benzoate-d4, Methyl p-(Methoxycarbonyl)benzoate-d4, NSC 3503-d4, 1,4-Benzene-2,3,5,6-d4-dicarboxylic acid, dimethyl ester (9CI), 1,4-Di(methoxycarbonyl)benzene-2,3,5,6-d4, 1,4-Dimethyl benzene-2,3,5,6-d4-1,4-dicarboxylate, Terephthalic acid dimethyl ester-d4, Terephthalic-d4 acid dimethyl ester, Terephthalic acid dimethyl ester D4 (phenyl-2,3,5,6-D4), Dimethyl terephthalate-2,3,5,6-D4
InChI formel InChI=1S/C10H10O4/c1-13-9(11)7-3-5-8(6-4-7)10(12)14-2/h3-6H,1-2H3/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)OC)c([2H])c([2H])c1C(=O)OC
IUPAC navn dimethyl 2,3,5,6-tetradeuteriobenzene-1,4-dicarboxylate
Molekylvægt (g/mol) 198.21
Formel vægt 198.083
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.