Få mere at vide
1,4-Benzenediamine-d8, CDN
Artikelnummer. 30323767
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30323767 0.25 g -
30311244 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323767 Leverandør CDN Leverandørnr. CDND33940.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,4-Phenylenediamine-d8
CAS 45619-88-3
Molekylær formel C6 D8 N2
Mængde 0.25 g
Synonym 1,4-Benzene-2,3,5,6-d4-di(amine-d2) (9 CI), 1,4-Phenylenediamine D8, 1,4-Phenylenediamine-d8, Benzene-1,4-diamine-d8, Perdeuterobenzene-1,4-diamine
InChI formel InChI=1 S/C6H8N2/c7-5-1-2-6(8)4-3-5/h1-4 H,7-8H2/i1D,2 D,3 D,4 D/hD4
SMIL [2 H]N([2 H])c1c([2 H])c([2 H])c(N([2 H])[2 H])c([2 H])c1[2 H]
IUPAC navn 1-N,1-N,4-N,4-N,2,3,5,6-octadeuteriobenzene-1,4-diamine
Molekylvægt (g/mol) 116.1904
Formel vægt 116.119 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity