missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(4-Acetylpiperidino)ethan-1-one, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 169.22 |
Percent Purity | 97% |
Chemical Name or Material | 1-(4-Acetylpiperidino)ethan-1-one |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'