Learn More
1,4,8,11-Tetrakis(aminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane, Thermo Scientific™
Catalog Number 15437585
Click to view available options
Quantity:
250 mg
1 g
345612-63-7
C18H36N8O4
428.54
MFCD09263308
IELGXFGSUVHRIY-UHFFFAOYSA-N
1,4,8,11-tetrakis aminocarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, tetam, acmc-20ap3g, cyclam-n,n',n,n'-tetraacetamide, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetamide, 1,4,8,11-tetrakis carbamoylmethyl-1,4,8,11-tetraazacyclotetradecane, 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetraacetamide, 2-4,8,11-tris carbamoylmethyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetamide
30682562
2-[4,8,11-tris(2-amino-2-oxoethyl)-1,4,8,11-tetrazacyclotetradec-1-yl]acetamide
NC(=O)CN1CCCN(CC(N)=O)CCN(CC(N)=O)CCCN(CC(N)=O)CC1
345612-63-7
C18H36N8O4
428.54
MFCD09263308
IELGXFGSUVHRIY-UHFFFAOYSA-N
1,4,8,11-tetrakis aminocarbonylmethyl-1,4,8,11-tetraazacyclotetradecane, tetam, acmc-20ap3g, cyclam-n,n',n,n'-tetraacetamide, 1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetraacetamide, 1,4,8,11-tetrakis carbamoylmethyl-1,4,8,11-tetraazacyclotetradecane, 2,2',2,2'-1,4,8,11-tetraazacyclotetradecane-1,4,8,11-tetrayl tetraacetamide, 2-4,8,11-tris carbamoylmethyl-1,4,8,11-tetraazacyclotetradecan-1-yl acetamide
30682562
2-[4,8,11-tris(2-amino-2-oxoethyl)-1,4,8,11-tetrazacyclotetradec-1-yl]acetamide
NC(=O)CN1CCCN(CC(N)=O)CCN(CC(N)=O)CCCN(CC(N)=O)CC1
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 428.54
Chemical Name or Material 1,4,8,11-Tetrakis(aminocarbonylmethyl)-1,4,8,11-tetraazacyclotetradecane