Learn More
1,4,7-Triazonane, TRC
Product Code. 30448941
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30448941 500 mg -
30457358 50 mg -
30455573 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30448941 Supplier Toronto Research Chemicals Supplier No. TRCT795273500MG
 Restricted Item
Forventet forsendelse: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 4730-54-5
Molecular Formula C6H15N3
Molecular Weight (g/mol) 129.2
IUPAC Name 1,4,7-triazonane
SMILES C1CNCCNCCN1
Tekniske data
Quantity 500 mg
InChI Formula InChI=1S/C6H15N3/c1-2-8-5-6-9-4-3-7-1/h7-9H,1-6H2
Formula Weight 129.1266
Chemical Name or Material 1,4,7-Triazonane