missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
1,4,5-Oxadiazepane Dihydrobromide, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 1,4,5-Oxadiazepane Dihydrobromide |
| CAS | 243973-70-8 |
| Molekylær formel | C4H10N2O . 2(HBr) |
| Mængde | 100 mg |
| Synonym | Hexahydro-1,4,5-oxadiazepine Dihydrobromide |
| InChI formel | InChI=1S/C4H10N2O.2BrH/c1-3-7-4-2-6-5-1;;/h5-6H,1-4H2;2*1H |
| SMIL | Br.O1CCNNCC1.Br |
| IUPAC navn | 1,4,5-oxadiazepane dihydrobromide |
| Molekylvægt (g/mol) | 263.96 |
| Formel vægt | 261.94 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?