Få mere at vide
1-[4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-1H-indol-1-yl]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol™
Artikelnummer. 30458173
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30458173 4 x 25 mg
30051881 25 mg
30051425 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30458173 Leverandør Mikromol Leverandørnr. MM0082.0800254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-[4-[2-Hydroxy-3-[(1-methylethyl)amino]propoxy]-1H-indol-1-yl]-3-[(1-methylethyl)amino]propan-2-ol, Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[4-[2-Hydroxy-3-[(1-methyl-ethyl)amino]propoxy]-1H-indol-1-yl]-3-[(1-methylethyl)amino]propan-2-ol
CAS 130115-63-8
Analyt- eller komponentnavne 1-[4-[2-Hydroxy-3-[(1-methyl-ethyl)amino]propoxy]-1H-indol-1-yl]-3-[(1-methylethyl)amino]propan-2-ol
Forsendelsestilstand Room Temperature
Molekylær formel C20 H33 N3 O3
Certificeringer/compliance ISO 17025
Synonym 1-[4-[2-Hydroxy-3-[(1-methyl-ethyl)amino]propoxy]-1H-indol-1-yl]-3-[(1-methylethyl)amino]propan-2-ol, Pindolol Imp. B (EP), Pindolol Impurity B
InChI formel InChI=1S/C20H33N3O3/c1-14(2)21-10-16(24)12-23-9-8-18-19(23)6-5-7-20(18)26-13-17(25)11-22-15(3)4/h5-9,14-17,21-22,24-25H,10-13H2,1-4H3
Løsningstype Neat
SMIL CC(C)NCC(O)COc1cccc2c1ccn2CC(O)CNC(C)C
IUPAC navn 1-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]indol-1-yl]-3-(propan-2-ylamino)propan-2-ol
Molekylvægt (g/mol) 363.49
Mængde 4 x 25 mg
Formel vægt 363.2522 g/mol