Få mere at vide
1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one, Mikromol™
Artikelnummer. 17971606
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17971606 100 mg
17983486 25 mg
30459389 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17971606 Leverandør Mikromol Leverandørnr. MM0468.05
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-[4-(1,1-dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one
CAS 97928-18-2
Analyt- eller komponentnavne 1-[4-(1,1-dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one
Forsendelsestilstand Room Temperature
Molekylær formel C19 H29 N O2
Certificeringer/compliance ISO 17025
Synonym Ebastine Imp. D (EP), 1-[4-(1,1-Dimethylethyl)phenyl]-4-(4-hydroxypiperidin-1-yl)butan-1-one, Ebastine Related Compound D, Ebastine Impurity D
InChI formel InChI=1S/C19H29NO2/c1-19(2,3)16-8-6-15(7-9-16)18(22)5-4-12-20-13-10-17(21)11-14-20/h6-9,17,21H,4-5,10-14H2,1-3H3
SMIL CC(C)(C)c1ccc(cc1)C(=O)CCCN2CCC(O)CC2
IUPAC navn 1-(4-tert-butylphenyl)-4-(4-hydroxypiperidin-1-yl)butan-1-one
Molekylvægt (g/mol) 303.44
Mængde 100 mg
Formel vægt 303.2198 g/mol