missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol, 98%, Thermo Scientific Chemicals

Catalog Number p-7026289
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
88462-65-1
C11H9F3O2
230.19
MFCD02683085
OPWHACCQYXGLRX-UHFFFAOYNA-N
1-3-trifluoromethylphenoxy-3-butyn-2-ol, 1-3-trifluoromethyl phenoxy but-3-yn-2-ol, 3-butyn-2-ol,1-3-trifluoromethyl phenoxy, acmc-20drht, 1-3-trifluoromethylphenoxy-3-butyn-2-o
11172256
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol
OC(COC1=CC=CC(=C1)C(F)(F)F)C#C
This item is not returnable. View return policy
88462-65-1
C11H9F3O2
230.19
MFCD02683085
OPWHACCQYXGLRX-UHFFFAOYNA-N
1-3-trifluoromethylphenoxy-3-butyn-2-ol, 1-3-trifluoromethyl phenoxy but-3-yn-2-ol, 3-butyn-2-ol,1-3-trifluoromethyl phenoxy, acmc-20drht, 1-3-trifluoromethylphenoxy-3-butyn-2-o
11172256
1-[3-(trifluoromethyl)phenoxy]but-3-yn-2-ol
OC(COC1=CC=CC(=C1)C(F)(F)F)C#C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30°C to 33°C
Quantity 1 g
Formula Weight 230.19
Percent Purity 98%
Chemical Name or Material 1-(3-Trifluoromethylphenoxy)-3-butyn-2-ol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'