Få mere at vide
1-(3-Pyridyl)-1,4-butanediol, TRC
Artikelnummer. 30006925
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30006925 10 mg
30003317 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30006925 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99250010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76014-83-0
Molekylær formel C9 H13 N O2
Molekylvægt (g/mol) 167.2
Synonym 1,4-Butanediol, 1-(3-pyridinyl)-, 1-(3-Pyridinyl)-1,4-butanediol, 1-(3-Pyridyl)-1,4-butanediol, 4-Hydroxy-4-(3-pyridyl)-butan-1-ol, 4-Hydroxy-4-(3-pyridyl)-butanol
IUPAC navn 1-pyridin-3-ylbutane-1,4-diol
SMIL OCCCC(O)c1cccnc1
Tekniske data
Kemisk navn eller materiale 1-(3-Pyridyl)-1,4-butanediol
Mængde 10 mg
InChI formel InChI=1S/C9H13NO2/c11-6-2-4-9(12)8-3-1-5-10-7-8/h1,3,5,7,9,11-12H,2,4,6H2
Formel vægt 167.0946