Få mere at vide
1,3-Propanediol-d6, TRC
Artikelnummer. 30420266
Skift visning
Mængde:
500 mg
Artikelnummer. Mængde
30420266 500 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30420266 Leverandør Toronto Research Chemicals Leverandørnr. TRCP760322500MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Propanediol-d6
CAS 284474-77-7
Molekylær formel C3H2D6O2
Mængde 500 mg
Synonym 1,3-Dihydroxypropane-d6, 1,3-Propylene Glycol-d6, 1,3-Propylenediol-d6, 2-Deoxyglycerol, Bio-PDO, NSC 65426, PG, Susterra, Trimethylene glycol, Zemea, Zemea propanediol, β-Propylene glycol, ω-Propanediol
InChI formel InChI=1S/C3H8O2/c4-2-1-3-5/h4-5H,1-3H2/i1D2,2D2,3D2
SMIL OC([2H])([2H])C([2H])([2H])C([2H])([2H])O
IUPAC navn propane-d6-1,3-diol
Molekylvægt (g/mol) 82.13
Formel vægt 82.09