missing translation for 'onlineSavingsMsg'
Learn More

1,3-Propane sultone, 97%, Thermo Scientific Chemicals

Catalog Number 15374968
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
1120-71-4
C3H6O3S
122.14
MFCD00005355
FSSPGSAQUIYDCN-UHFFFAOYSA-N
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
14264
CHEBI:82370
oxathiolane 2,2-dioxide
O=S1(=O)CCCO1
This item is not returnable. View return policy
1120-71-4
C3H6O3S
122.14
MFCD00005355
FSSPGSAQUIYDCN-UHFFFAOYSA-N
1,3-propanesultone, propane sultone, 1,2-oxathiolane 2,2-dioxide, 1,3-propane sultone, 1,2-oxathiolane, 2,2-dioxide, propanesultone, rcra waste number u193, .gamma.-propane sultone, 3-hydroxy-1-propanesulfonic acid gamma-sultone, unii-l6ntk7vjx9
14264
CHEBI:82370
oxathiolane 2,2-dioxide
O=S1(=O)CCCO1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30.0°C to 33.0°C
Density 1.3920g/mL
Boiling Point 180.0°C (30.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 1 kg
Specific Gravity 1.392
Formula Weight 122.14
Percent Purity 97%
Chemical Name or Material 1,3-Propane sultone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'