Få mere at vide
1,3-Propane-2,2-d2-diol, CDN
Artikelnummer. 30322403
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322403 0.1 g 0.1g
30322191 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322403 Leverandør CDN Leverandørnr. CDND56510.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Propanediol-2,2-D2
CAS 38645-14-6
Molekylær formel C3 D2 H6 O2
Mængde 0.1 g
Synonym 1,3-Propane-2,2-d2-diol (9 CI), 1,3-Propanediol-2,2-d2, 1,3-Propanediol-2,2-D2, 1,3-Propanediol-d2
InChI formel InChI=1 S/C3H8O2/c4-2-1-3-5/h4-5 H,1-3H2/i1D2
SMIL [2 H]C([2 H])(CO)CO
IUPAC navn 2,2-dideuteriopropane-1,3-diol
Molekylvægt (g/mol) 78.1067
Formel vægt 78.065 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity