Få mere at vide
1,3-Propane-2,2-d2-diamine 2HCl, CDN
Artikelnummer. 30320071
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30320071 0.05 g -
30314654 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320071 Leverandør CDN Leverandørnr. CDND51500.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Diaminopropane-2,2-D2 Dihydrochloride
CAS 352438-79-0
Molekylær formel C3 D2 H8 N2 . 2 Cl H
Mængde 0.05 g
Synonym 1,3-Propane-2,2-d2-diamine, dihydrochloride (9 CI), 2,2-Dideuteriopropane-1,3-diamine dihydrochloride, 1,3-Diaminopropane-d2 dihydrochloride, 1,3-Diaminopropane-2,2-D2 dihydrochloride, Trimethylenediamine-d2 dihydrochloride
InChI formel InChI=1 S/C3H10N2.2 ClH/c4-2-1-3-5;;/h1-5H2;2*1 H/i1D2;;
SMIL Cl.Cl.[2 H]C([2 H])(CN)CN
IUPAC navn 2,2-dideuteriopropane-1,3-diamine;dihydrochloride
Molekylvægt (g/mol) 149.0591
Formel vægt 148.0503 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity