Få mere at vide
1,3-Propane-1,1,3,3-d4-diamine, CDN
Artikelnummer. 30318124
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318124 0.05 g
30309452 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318124 Leverandør CDN Leverandørnr. CDND37310.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Diaminopropane-1,1,3,3-D4
CAS 347840-13-5
Molekylær formel C3 D4 H6 N2
Mængde 0.05 g
Synonym 1,3-Propane-1,1,3,3-d4-diamine (9 CI), 1,1,3,3-Tetradeuteriopropane-1,3-diamine, 1,3-Diaminopropane-d4, 1,3-Diaminopropane-1,1,3,3-D4, Trimethylenediamine-d4
InChI formel InChI=1 S/C3H10N2/c4-2-1-3-5/h1-5H2/i2D2,3D2
SMIL [2 H]C([2 H])(N)CC([2 H])([2 H])N
IUPAC navn 1,1,3,3-tetradeuteriopropane-1,3-diamine
Molekylvægt (g/mol) 78.1496
Formel vægt 78.1095 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity