Få mere at vide
1,3-Phenylenediamine-d4, TRC
Artikelnummer. 30410426
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30410426 10 mg
30425716 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30410426 Leverandør Toronto Research Chemicals Leverandørnr. TRCP31983710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 770735-58-5
Molekylær formel C6 D7 H N2
Molekylvægt (g/mol) 115.1843
Synonym 1,3-Benzene-2,4,5,6-d4-di(amine-d2) (9CI), 1,3-Benzenediamine-d8, m-Phenylenediamine-d8, 1,3-Diaminobenzene-d8, 1,3-Phenylenediamine-d8, 3-Aminoaniline-d8, m-Diaminobenzene-d8, 1,3-Phenylenediamine-D8
IUPAC navn 1-N,1-N,3-N,2,4,5,6-heptadeuteriobenzene-1,3-diamine
SMIL [2H]Nc1c([2H])c([2H])c([2H])c(N([2H])[2H])c1[2H]
Tekniske data
Kemisk navn eller materiale 1,3-Phenylenediamine-d4
Mængde 10 mg
InChI formel InChI=1S/C6H8N2/c7-5-2-1-3-6(8)4-5/h1-4H,7-8H2/i1D,2D,3D,4D/hD3
Formel vægt 115.1127