Få mere at vide
1,3-O-Bis(triisopropylsilyl) 2-Oleoyl Glycerol-d5, TRC
Artikelnummer. 30334745
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30334745 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334745 Leverandør Toronto Research Chemicals Leverandørnr. TRCB58866250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-O-Bis(triisopropylsilyl) 2-Oleoyl Glycerol-d5
CAS 1331658-79-7
Molekylær formel C39H75D5O4Si2
Mængde 50 mg
Synonym (9Z)-9-Octadecenoic Acid 2-Triisopropylsilyloxy-1-(triisopropylsilyloxymethyl)ethyl-d5 Ester, 1,3-O-Bis(triisopropylsilyl)-2-monoolein-d5, 1,3-O-Bis(triisopropylsilyl)-2-monooleoylglycerol-d5, 1,3-O-Bis(triisopropylsilyl)-2-oleoylglycerol-d5, 1,3-O-Bis(triisopropylsilyl)-glycerol-d5 1,3-O-Bis(triisopropylsilyl)-2-monooleate, 1,3-O-Bis(triisopropylsilyl)-beta-monoolein-d5
InChI formel InChI=1S/C39H80O4Si2/c1-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-39(40)43-38(30-41-44(32(2)3,33(4)5)34(6)7)31-42-45(35(8)9,36(10)11)37(12)13/h21-22,32-38H,14-20,23-31H2,1-13H3/b22-21-/i30D2,31D2,38D
SMIL [2H]C([2H])(O[Si](C(C)C)(C(C)C)C(C)C)C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])([2H])O[Si](C(C)C)(C(C)C)C(C)C
IUPAC navn [1,1,2,3,3-pentadeuterio-1,3-bis[tri(propan-2-yl)silyloxy]propan-2-yl] (Z)-octadec-9-enoate
Molekylvægt (g/mol) 674.25
Formel vægt 673.5909