Learn More
1-(3-Methoxyphenyl)ethan-1-one, Mikromol™
Product Code. 17920616
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity Unit Size
17920616 100 mg 100mg
1 options
Product Code. 17920616 Supplier Mikromol Supplier No. MM1497.220100
 Restricted Item
Forventet forsendelse: 26-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(3-Methoxyphenyl)ethan-1-one
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(3-Methoxyphenyl)ethan-1-one
CAS 586-37-8
Analyte or Component Names 1-(3-Methoxyphenyl)ethan-1-one
Shipping Condition Room Temperature
Molecular Formula C9 H10 O2
Certifications/Compliance ISO 9001
Synonym Ethanone, 1-(3-methoxyphenyl)-, Acetophenone, 3'-methoxy- (8CI), 1-(3-Methoxyphenyl)ethanone, 1-Acetyl-3-methoxybenzene, 3-Acetylanisole, 3-Methoxy-1-acetylbenzene, 3-Methoxyacetophenone, 3-Methoxyphenyl methyl ketone, 3'-Methoxyacetophenon, 3'-Methoxyacetophenone, Methyl 3-methoxyphenyl ketone, NSC 65593, m-Acetylanisole, m-Methoxyacetophenone, Rivastigmine Hydrogen Tartrate Imp. H (EP), Rivastigmine Hydrogen Tartrate Impurity H, Rivastigmine Impurity H
InChI Formula InChI=1S/C9H10O2/c1-7(10)8-4-3-5-9(6-8)11-2/h3-6H,1-2H3
SMILES COc1cccc(c1)C(=O)C
IUPAC Name 1-(3-methoxyphenyl)ethanone
Molecular Weight (g/mol) 150.17
Quantity 100 mg
Formula Weight 150.0681 g/mol