missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorophenyl)pyrrole, 98%, Thermo Scientific™

Catalog Number p-7032030
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
53871-27-5
C10H8FN
161.18
MFCD02665249
MMILNBLSOHVLCJ-UHFFFAOYSA-N
1-3-fluorophenyl pyrrole, 1-3-fluorophenyl-1h-pyrrole
4763720
1-(3-fluorophenyl)pyrrole
FC1=CC=CC(=C1)N1C=CC=C1
This item is not returnable. View return policy
53871-27-5
C10H8FN
161.18
MFCD02665249
MMILNBLSOHVLCJ-UHFFFAOYSA-N
1-3-fluorophenyl pyrrole, 1-3-fluorophenyl-1h-pyrrole
4763720
1-(3-fluorophenyl)pyrrole
FC1=CC=CC(=C1)N1C=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Boiling Point 54°C (0.15 mmHg)
Refractive Index 1.578
Quantity 1 g
Formula Weight 161.18
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)pyrrole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'