missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorobenzyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog Number p-7052683
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
55513-19-4
C11H15FN2
194.253
MFCD02242870
ITHBJSRWFNLKIH-UHFFFAOYSA-N
1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate
903813
1-[(3-fluorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC(=CC=C2)F
This item is not returnable. View return policy
55513-19-4
C11H15FN2
194.253
MFCD02242870
ITHBJSRWFNLKIH-UHFFFAOYSA-N
1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate
903813
1-[(3-fluorophenyl)methyl]piperazine
C1CN(CCN1)CC2=CC(=CC=C2)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Orange
Density 1.106 g/mL
Boiling Point 281°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Formula Weight 194.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3-Fluorobenzyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'