Få mere at vide
1,3-Distearoyl-2-oleoyl Glycerol, TRC
Artikelnummer. 30003931
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003931 5 mg
30004494 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30003931 Leverandør Toronto Research Chemicals Leverandørnr. TRCD4935255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2846-04-0
Molekylær formel C57 H108 O6
Molekylvægt (g/mol) 889.46
Synonym 1,3-Di-O-stearoyl-2-O-oleoylglycerol, 1,3-Distearo-2-olein, 1,3-Distearoyl-2-olein, 1,3-Distearoyl-2-oleoylglycerol, 1,3-Distearyl-2-oleylglycerol, 2-Oleo-1,3-distearin, 2-Oleoyl-1,3-distearin, 2-Oleoyl-1,3-distearoylglycerol, Chocoseed, Glyceryl 2-Oleoyl-1,3-distearate, SOS, Triglyceride StOSt,sn
IUPAC navn [3-octadecanoyloxy-2-[(E)-octadec-9-enoyl]oxy-propyl] octadecanoate
SMIL CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)OC(=O)CCCCCCC\C=C\CCCCCCCC
Tekniske data
Kemisk navn eller materiale 1,3-Distearoyl-2-oleoyl Glycerol
Mængde 5 mg
InChI formel InChI=1S/C57H108O6/c1-4-7-10-13-16-19-22-25-28-31-34-37-40-43-46-49-55(58)61-52-54(63-57(60)51-48-45-42-39-36-33-30-27-24-21-18-15-12-9-6-3)53-62-56(59)50-47-44-41-38-35-32-29-26-23-20-17-14-11-8-5-2/h27,30,54H,4-26,28-29,31-53H2,1-3H3/b30-27+
Formel vægt 888.8146