missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3-Diphenylisobenzofuran, 95%, BLD Pharm™

Product Code. 31001349
Encompass_Preferred
Change view
Click to view available options
Mængde:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde Unit Size
31001349 100 g 100g
31000607 5 g 5g
30988845 1 g 1g
30996884 10 g 10g
30996063 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31001349 Supplier BLD Pharm Supplier No. BD1137100g
 Begrænset vare
Forventet forsendelse: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Kemisk navn eller materiale 1,3-Diphenylisobenzofuran
CAS 5471-63-6
Molekylær formel C20H14O
MDL nummer MFCD00005931
Mængde 100 g
SMIL C1(C2=CC=CC=C2)=C3C=CC=CC3=C(C4=CC=CC=C4)O1
Molekylvægt (g/mol) 270.32
Procent renhed 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.