missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
1,3-Dipalmitoyl-2-chloropropanediol D5 (2-chloro-1,3-propanediol D5) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | 1,3-Dipalmitoyl-2-chloropropanediol-d5 |
| CAS | 1426395-62-1 |
| Opløsningsmiddel eller Matrix | Acetonitrile |
| Molekylær formel | C35 2H5 H62 Cl O4 |
| Mængde | 1 mL |
| Synonym | 1,3-Dipalmitoyl-2-chloropropanediol D5 (2-Chloro-1,3-propanediol D5), Hexadecanoic acid, 1,1'-(2-chloro-1,3-propanediyl-1,1,2,3,3-d5) ester |
| InChI formel | InChI=1S/C35H67ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-31-33(36)32-40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33H,3-32H2,1-2H3/i31D2,32D2,33D |
| SMIL | [2H]C([2H])(OC(=O)CCCCCCCCCCCCCCC)C([2H])(Cl)C([2H])([2H])OC(=O)CCCCCCCCCCCCCCC |
| IUPAC navn | (2-chloro-1,1,2,3,3-pentadeuterio-3-hexadecanoyloxypropyl) hexadecanoate |
| Molekylvægt (g/mol) | 592.39 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?