missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5%, stab., Thermo Scientific Chemicals

Catalog Number p-7058620
Click to view available options
Quantity:
250 mL
1 L
4 x 1 L
646-06-0
C3H6O2
74.079
MFCD00003207
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
12586
CHEBI:87597
1,3-dioxolane
C1COCO1
646-06-0
C3H6O2
74.079
MFCD00003207
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
12586
CHEBI:87597
1,3-dioxolane
C1COCO1

As a solvent, in Mannich reaction, as an electrolyte in batteries

As a solvent, in Mannich reaction, as an electrolyte in batteries.1,3-Dioxolane is used as a solvent for paints, cellulose esters and in organic synthesis. It is used as an extragent for waxes, fats and oils, stabilizer for halogenated hydrocarbons and co monomer in polyacetals preparation. It is used as finishing agent and swelling agent in textiles. It acts as an intermediate for the preparation of acyclovir and vandetanib. It is an essential ingredient in industrial polymers. In polymerization reactions, it acts as a chain length regulator and a chain transfer agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -95°C
Density 1.06 g/mL
Boiling Point 74°C to 75°C
Flash Point 1°C (34°F)
Refractive Index 1.4
Quantity 250 mL
UN Number UN1166
Solubility Information Miscible with water,ethanol,ether,n-octanol,acetone,chloroform,dichloromethane,ethyl Acetate and methanol.
Formula Weight 74.08
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material 1,3-Dioxolane, Stabilized
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'