Få mere at vide
1,3-Dinitrobenzene-d4, CDN
Artikelnummer. 30321675
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30321675 5 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321675 Leverandør CDN Leverandørnr. CDND19375G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dinitrobenzene-d4
CAS 54247-05-1
Molekylær formel C6 2H4 N2 O4
Mængde 5 g
Synonym Benzene-1,2,3,5-d4, 4,6-dinitro-, Benzene-1,2,3,5-d4, dinitro- (6 CI), 4,6-Dinitrobenzene-1,2,3,5-d4, m-Dinitrobenzene-d4, 1,3-Dinitrobenzene D4
InChI formel InChI=1 S/C6H4N2O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(c([2 H])c(c1[2 H])[N+](=O)[O-])[N+](=O)[O-]
IUPAC navn 1,2,3,5-tetradeuterio-4,6-dinitrobenzene
Molekylvægt (g/mol) 172.13
Formel vægt 172.0422 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity