Learn More
1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, Sigma-Aldrich™
Product Code. 15890301
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15890301 100 mL 100mL
15800311 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15890301 Supplier Sigma Aldrich Supplier No. 8182140100
 Restricted Item
Forventet forsendelse: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7226-23-5
Grade Research
Molecular Weight (g/mol) 128.17
Specifications
Melting Point -24°C
Density 1.064 g/cm3 at 20°C
Boiling Point 247°C/1013 hPa
MDL Number MFCD00006550
Quantity 100 mL
Synonym 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone, Dimethylpropyleneurea, DMPU, 1,3-Dimethyltetrahydro-2(1H)-pyrimidinone