Få mere at vide
1,3-Dimethylpentylamine Hydrochloride Salt, TRC
Artikelnummer. 30132567
Skift visning
Mængde:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132567 50 mg
30132430 500 mg
17984115 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132567 Leverandør Toronto Research Chemicals Leverandørnr. TRCD47660050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 13803-74-2
Molekylær formel C7 H17 N . Cl H
Molekylvægt (g/mol) 151.68
Synonym Dimethylamylamine Hydrochloride, 4-Methyl-2-hexanamine Hydrochloride, DMAA HCl
IUPAC navn 4-methylhexan-2-amine;hydrochloride
SMIL Cl.CCC(C)CC(C)N
Tekniske data
Kemisk navn eller materiale Methylhexanamine hydrochloride
Mængde 50 mg
InChI formel InChI=1S/C7H17N.ClH/c1-4-6(2)5-7(3)8;/h6-7H,4-5,8H2,1-3H3;1H
Formel vægt 151.1128