missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3-Dimethylnaphthalene, SPEX CertiPrep™

Artikelnummer. 12999194 Shop alle SPEX Certiprep produkter
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 mL
Pakningsstørrelse:
1ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde Pakningsstørrelse
12999194 1 mL 1ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 12999194 Leverandør SPEX Certiprep Leverandørnr. S1631

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,3-Dimethylnaphthalene575-41-71000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Kemisk navn eller materiale 1,3-Dimethylnaphthalene
Emballage 2mL amber prescored glass ampule
CAS 575-41-7, 67-56-1
CAS min % 0.1, 99.9
Koncentration eller sammensætningsnoter Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
CAS Max % 0.1, 99.9
Molekylær formel C12H12
MDL nummer MFCD00004036
Til brug med (udstyr) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
InChI nøgle QHJMFSMPSZREIF-UHFFFAOYSA-N
SMIL CC1=CC(C)=C2C=CC=CC2=C1
IUPAC navn 1,3-dimethylnaphthalene
Opløsningsmiddel eller Matrix Methanol
Molekylvægt (g/mol) 156.23
PubChem CID 11327
Mængde 1 mL
ChEBI CHEBI:48614
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.