Learn More
1,3-dimethyl-1h-pyrazole-4-boronic acid,pinacol ester, 97%, Thermo Scientific™
Product Code. 10235324
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
Product Code. Quantity Packaging Unit Size
10235324 1 g Amber Glass Bottle 1g
1 options
Product Code. 10235324 Supplier Thermo Scientific Maybridge Supplier No. MAY00075DA
 Restricted Item
Forventet forsendelse: 05-02-2027
Specifications
CAS 1046832-21-6
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H19BN2O2
MDL Number MFCD09864190
Quantity 1 g
Synonym 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3-dimethyl-1h-pyrazole-4-boronic acid,pinacol ester, 1,3-dimethylpyrazole-4-boronic acid pinacol ester, 1,3-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1,3-dimethyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 2-1,3-dimethylpyrazol-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyrazole, 1h-pyrazole, 1,3-dimethyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, pubchem23961
InChI Key FBNAMBTYMSWTIB-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2C)C
IUPAC Name 1,3-dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
Molecular Weight (g/mol) 222.095
PubChem CID 42647316
Formula Weight 222.09
Percent Purity 97%
Chemical Name or Material 1, 3-Dimethyl-1H-pyrazole-4-boronic acid, pinacol ester