Få mere at vide
1,3-Dimethoxybenzene-2,4,5,6-d4, CDN
Artikelnummer. 30322051
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30322051 0.25 g -
30309260 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322051 Leverandør CDN Leverandørnr. CDND54290.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dimethoxybenzene-2,4,5,6-D4
CAS 362049-44-3
Molekylær formel C8 D4 H6 O2
Mængde 0.25 g
Synonym Benzene-1,2,3,5-d4, 4,6-dimethoxy- (9 CI), 1,3-Dimethoxybenzene-2,4,5,6-D4, 1,3-Dimethoxybenzene-d4, 3-Methoxyanisole-d4, 3-Methoxyphenyl-d4 methyl ether, Resorcinol-d4 dimethyl ether, m-Dimethoxybenzene-d4, m-Methoxyanisole-d4
InChI formel InChI=1 S/C8H10O2/c1-9-7-4-3-5-8(6-7)10-2/h3-6 H,1-2H3/i3D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c(OC)c([2 H])c(OC)c1[2 H]
IUPAC navn 1,2,3,5-tetradeuterio-4,6-dimethoxybenzene
Molekylvægt (g/mol) 142.188
Formel vægt 142.093 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity