Få mere at vide
1,3-Dimethoxy-d6-benzene, CDN
Artikelnummer. 30317911
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317911 0.5 g 0.5g
30314641 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317911 Leverandør CDN Leverandørnr. CDND54300.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dimethoxybenzene-D6 (dimethoxy-D6)
CAS 16469-85-5
Molekylær formel C8 D6 H4 O2
Mængde 0.5 g
Synonym Benzene, 1,3-di(methoxy-d3)- (9 CI), Benzene, m-di(methoxy-d3)- (8 CI), 1,3-Dimethoxybenzene-D6 (dimethoxy-D6), 1,3-Dimethoxybenzene-d6, 3-Methoxyanisole-d6, 3-(Methoxy-d3)phenyl methyl-d3 ether, Resorcinol di(methyl-d3) ether, m-Dimethoxybenzene-d6, m-Methoxyanisole-d6
InChI formel InChI=1 S/C8H10O2/c1-9-7-4-3-5-8(6-7)10-2/h3-6 H,1-2H3/i1D3,2D3
SMIL [2 H]C([2 H])([2 H])Oc1cccc(OC([2 H])([2 H])[2 H])c1
IUPAC navn 1,3-bis(trideuteriomethoxy)benzene
Molekylvægt (g/mol) 144.201
Formel vægt 144.106 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity