missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dihydro-1,3-dioxo-5-Isobenzofurancarboxylic Acid 2-Ethylhexyl Ester, TRC
Specifications
Specifications
| Kemisk navn eller materiale | 1,3-Dihydro-1,3-dioxo-5-isobenzofurancarboxylic Acid |
| CAS | 26597-44-4 |
| Molekylær formel | C17H20O5 |
| Mængde | 250 mg |
| Synonym | 1,3-Dihydro-1,3-Dioxo-5-Isobenzofurancarboxylic Acid |
| InChI formel | InChI=1S/C17H20O5/c1-3-5-6-11(4-2)10-21-15(18)12-7-8-13-14(9-12)17(20)22-16(13)19/h7-9,11H,3-6,10H2,1-2H3 |
| SMIL | O=C1OC(C2=CC(C(OCC(CC)CCCC)=O)=CC=C21)=O |
| IUPAC navn | 2-ethylhexyl 1,3-dioxo-1,3-dihydroisobenzofuran-5-carboxylate |
| Molekylvægt (g/mol) | 304.34 |
| Formel vægt | 304.13 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?