Få mere at vide
1,3-Difluorobenzene-d4, CDN
Artikelnummer. 30318400
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30318400 0.25 g -
30317648 0.1 g 0.1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318400 Leverandør CDN Leverandørnr. CDND76120.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Difluorobenzene-d4
CAS 1173022-08-6
Molekylær formel C6 2H4 F2
Mængde 0.25 g
InChI formel InChI=1 S/C6H4F2/c7-5-2-1-3-6(8)4-5/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(F)c([2 H])c(F)c1[2 H]
IUPAC navn 1,2,3,5-tetradeuterio-4,6-difluorobenzene
Molekylvægt (g/mol) 118.12
Formel vægt 118.0532 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity