missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoro-5-nitrobenzene, 98%, Thermo Scientific™

Catalog Number 15446748
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2265-94-3
C6H3F2NO2
159.09
MFCD00012142
AUQBBDWDLJSKMI-UHFFFAOYSA-N
3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
75272
1,3-difluoro-5-nitrobenzene
[O-][N+](=O)C1=CC(F)=CC(F)=C1
This item is not returnable. View return policy
2265-94-3
C6H3F2NO2
159.09
MFCD00012142
AUQBBDWDLJSKMI-UHFFFAOYSA-N
3,5-difluoronitrobenzene, benzene, 1,3-difluoro-5-nitro, 1,3-difluoro-5-nitro-benzene, 3,5-difluoro-1-nitrobenzene, 1-nitro-3,5-difluorobenzene, benzene, 1,3-difluoro-5-nitro-7ci,8ci,9ci, 3,5-difluoronitroben, pubchem2291, 3,5-difluornitrobenzene, acmc-1ck2x
75272
1,3-difluoro-5-nitrobenzene
[O-][N+](=O)C1=CC(F)=CC(F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 17°C
Density 1.407
Boiling Point 176°C to 177°C
Flash Point 74°C (165°F)
Refractive Index 1.499
Quantity 1 g
Formula Weight 159.09
Percent Purity 98%
Chemical Name or Material 1,3-Difluoro-5-nitrobenzene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'