Få mere at vide
1,3-Diethylbenzene-2,4,5,6-d4, CDN
Artikelnummer. 30319922
Skift visning
Mængde:
0.1 g
0.25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319922 0.1 g
30314512 0.25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319922 Leverandør CDN Leverandørnr. CDND63070.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Diethylbenzene-2,4,5,6-D4
CAS 1219803-29-8
Molekylær formel C10 D4 H10
Mængde 0.1 g
Synonym 1,3-Diethylbenzene-2,4,5,6-d4, m-Diethylbenzene-d4
InChI formel InChI=1 S/C10H14/c1-3-9-6-5-7-10(4-2)8-9/h5-8 H,3-4H2,1-2H3/i5D,6 D,7 D,8 D
SMIL [2 H]c1c([2 H])c(CC)c([2 H])c(CC)c1[2 H]
IUPAC navn 1,2,3,5-tetradeuterio-4,6-diethylbenzene
Molekylvægt (g/mol) 138.2428
Formel vægt 138.1347 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity