Få mere at vide
(1,3-Dichloropropyl)benzene, TRC
Artikelnummer. 30366332
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30366332 500 mg
30368485 1 g
30353058 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30366332 Leverandør Toronto Research Chemicals Leverandørnr. TRCD188470500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14155-36-3
Molekylær formel C9H10Cl2
Molekylvægt (g/mol) 189.08
Synonym 1,3-Dichloro-1-phenylpropane
IUPAC navn 1,3-dichloropropylbenzene
SMIL ClCCC(Cl)c1ccccc1
Tekniske data
Kemisk navn eller materiale (1,3-Dichloropropyl)benzene
Mængde 500 mg
InChI formel InChI=1S/C9H10Cl2/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5,9H,6-7H2
Formel vægt 188.016