Få mere at vide
1,3-Dichlorobenzene-d4, CDN
Artikelnummer. 30320681
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320681 0.5 g
30315420 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320681 Leverandør CDN Leverandørnr. CDND24050.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dichlorobenzene-d4
CAS 2199-70-4
Molekylær formel C6 2H4 Cl2
Mængde 0.5 g
Synonym Benzene-1,2,3,5-d4, 4,6-dichloro- (8 CI,9 CI), Benzene-1,2,3,5-d4, dichloro- (7 CI), 1,3-Dichlorobenzene D4, 1,3-Dichlorobenzene-d4
InChI formel InChI=1 S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4 H/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(Cl)c([2 H])c(Cl)c1[2 H]
IUPAC navn 1,3-dichloro-2,4,5,6-tetradeuteriobenzene
Molekylvægt (g/mol) 151.03
Formel vægt 149.9941 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity