Learn More
1,3-Dichloroacetone, 96%, ACROS Organics™
Catalog Number p-103920
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
534-07-6
C3H4Cl2O
126.96
MFCD00000937
SUNMBRGCANLOEG-UHFFFAOYSA-N
1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
10793
1,3-dichloropropan-2-one
ClCC(=O)CCl
534-07-6
C3H4Cl2O
126.96
MFCD00000937
SUNMBRGCANLOEG-UHFFFAOYSA-N
1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
10793
1,3-dichloropropan-2-one
ClCC(=O)CCl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White or Colorless
Density 1.3830g/mL
Boiling Point 172°C to 173°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCH2COCH2Cl
Quantity 25 g
Beilstein 01, 655
Fieser 02,109
Merck Index 15, 3061
Specific Gravity 1.383
Solubility Information Solubility in water: 27.9g/L water (20° c). Other solubilities: 464g/L toluene (20°C),soluble in alcohol,ether and hexane,slightly soluble in acetone,benzene,chloroform,and methanol
Formula Weight 126.97
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 1, 3-Dichloroacetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloroacetone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.