Få mere at vide
1,3-Dichloro-2-propanol-d5, TRC
Artikelnummer. 30373219
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30373219 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30373219 Leverandør Toronto Research Chemicals Leverandørnr. TRCD43580210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1,3-Dichloropropan-2-ol-d5
CAS 1173020-20-6
Renhedsgrad noter GC
Molekylær formel C3 D5 H Cl2 O
Mængde 10 mg
Synonym d5-1,3-Dichloro-2-propanol, 1,3-Dichloro-2-propanol-d5, 1,3-Dichloro-2-hydroxypropane-d5, 1,3-Dichlorohydrin-d5, 1,3-Dichloroisopropanol-d5, 1,3-Dichloroisopropyl Alcohol-d5, 2-Chloro-1-(chloromethyl)ethanol-d5, Bis(chloromethyl)methanol-d5, Glycerol 1,3-Dichlorohydrin-d5, NSC 70982, Propylene Dichlorohydrin
InChI formel InChI=1S/C3H6Cl2O/c4-1-3(6)2-5/h3,6H,1-2H2/i1D2,2D2,3D
SMIL [2H]C([2H])(Cl)C([2H])(O)C([2H])([2H])Cl
IUPAC navn 1,3-dichloro-1,1,2,3,3-pentadeuteriopropan-2-ol
Molekylvægt (g/mol) 134.02
Formel vægt 133.011
Procent renhed >95