missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromotetrafluorobenzene, 98+%, Thermo Scientific Chemicals

Catalog Number 11443637
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1559-87-1
C6Br2F4
307.87
MFCD00031533
UCWKDDQEZQRGDR-UHFFFAOYSA-N
73793
1,3-dibromo-2,4,5,6-tetrafluorobenzene
FC1=C(F)C(Br)=C(F)C(Br)=C1F
This item is not returnable. View return policy
1559-87-1
C6Br2F4
307.87
MFCD00031533
UCWKDDQEZQRGDR-UHFFFAOYSA-N
73793
1,3-dibromo-2,4,5,6-tetrafluorobenzene
FC1=C(F)C(Br)=C(F)C(Br)=C1F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 2.18
Boiling Point 85°C to 88°C (18 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.516
Quantity 1 g
Synonym 1,3-dibromotetrafluorobenzene, benzene, 1,3-dibromotetrafluoro, benzene, 1,3-dibromo-2,4,5,6-tetrafluoro, 1,3-dibromo-2,4,5,6-tetrafluoro-benzene, benzene,1,3-dibromo-2,4,5,6-tetrafluoro, acmc-1byn7, benzene,3-dibromotetrafluoro, 1,3-dibromotetrafluorobenzene, 98+%
Formula Weight 307.87
Percent Purity ≥98%
Chemical Name or Material 1,3-Dibromotetrafluorobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'