Learn More
1,3-Dibromopropane, Sigma-Aldrich™
Product Code. 15858990
Change view
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15858990 250 mL 250mL
15848990 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15858990 Supplier Sigma Aldrich Supplier No. 8032790250
 Restricted Item
Forventet forsendelse: 20-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 109-64-8
Grade Research
Molecular Formula Br(CH2)3Br
Molecular Weight (g/mol) 201.89
Specifications
Melting Point -34°C
Density 1.98 g/cm3 at 20°C
Boiling Point 166°C to 168°C/1013 hPa
MDL Number MFCD00000255
Quantity 250 mL
Synonym 1,3-Dibromopropane, Trimethylene bromide, 1,3-Dibromopropane