Learn More
Alfa Aesar™ 1,3-Dibromo-4-nitrobenzene, 98%
Brand: Alfa Aesar™ H61738.06
Additional Details : CAS Number : 51686-78-3 Weight : 0.00500kg Transport : UN number : 3459 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
51686-78-3 | |
280.903 | |
DXRVYZGVVFZCFP-UHFFFAOYSA-N | |
12315646 | |
C1=CC(=C(C=C1Br)Br)[N+](=O)[O-] |
C6H3Br2NO2 | |
MFCD02660416 | |
2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene | |
2,4-dibromo-1-nitrobenzene |
Specifications
1,3-Dibromo-4-nitrobenzene | |
5g | |
C6H3Br2NO2 | |
UN3459 | |
DXRVYZGVVFZCFP-UHFFFAOYSA-N | |
2,4-dibromo-1-nitrobenzene | |
12315646 | |
Crystalline Powder |
Yellow | |
51686-78-3 | |
MFCD02660416 | |
2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene | |
C1=CC(=C(C=C1Br)Br)[N+](=O)[O-] | |
280.903 | |
280.9 | |
98% |