missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3-Dibenzyloxy-2-propanol, TRC

Artikelnummer. 30377288
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30377288 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30377288 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41808050G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 1,3-Dibenzyloxy-2-propanol
CAS 6972-79-8
Molekylær formel C17 H20 O3
Mængde 50 g
InChI formel InChI=1S/C17H20O3/c18-17(13-19-11-15-7-3-1-4-8-15)14-20-12-16-9-5-2-6-10-16/h1-10,17-18H,11-14H2
SMIL OC(COCc1ccccc1)COCc2ccccc2
IUPAC navn 1,3-bis(phenylmethoxy)propan-2-ol
Molekylvægt (g/mol) 272.34
Formel vægt 272.1412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.