missing translation for 'onlineSavingsMsg'
Få mere at vide

1,3-Diaminopropane Dihydrochloride (Low water content), TRC

Artikelnummer. 30367559
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30367559 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30367559 Leverandør Toronto Research Chemicals Leverandørnr. TRCD41769510G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kræver en prisopdatering, inden den kan bestilles.

Anmod om en pris på denne vare her.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 1,3-Diaminopropane Dihydrochloride
CAS 10517-44-9
Molekylær formel C3H10N2 • 2HCl
Mængde 10 g
Synonym Trimethylenediamine Dihydrochloride
InChI formel InChI=1S/C3H10N2.2ClH/c4-2-1-3-5;;/h1-5H2;2*1H
SMIL NCCCN.Cl.Cl
IUPAC navn propane-1,3-diamine hydrochloride
Molekylvægt (g/mol) 147.04
Formel vægt 146.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.