Få mere at vide
1,3-Diacetoxypropane, TRC
Artikelnummer. 30363115
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30363115 250 mg -
30354681 2.5 g 2.5g
30348818 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363115 Leverandør Toronto Research Chemicals Leverandørnr. TRCD307573250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 628-66-0
Molekylær formel C7H12O4
Molekylvægt (g/mol) 160.17
IUPAC navn 3-acetyloxypropyl acetate
SMIL CC(=O)OCCCOC(=O)C
Tekniske data
Kemisk navn eller materiale 1,3-Diacetoxypropane
Mængde 250 mg
InChI formel InChI=1S/C7H12O4/c1-6(8)10-4-3-5-11-7(2)9/h3-5H2,1-2H3
Formel vægt 160.0736