Få mere at vide
1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride, TRC
Artikelnummer. 30352612
Skift visning
Mængde:
2 g
Artikelnummer. Mængde
30352612 2 g
1 options
Artikelnummer. 30352612 Leverandør Toronto Research Chemicals Leverandørnr. TRCC3797452G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 1-(3-Chloropropyl)-4-methylpiperazine Dihydrochloride
CAS 2031-23-4
Molekylær formel C8H19Cl3N2
Mængde 2 g
Synonym 1-(3-Chloropropyl)-4-methyl-piperazine Hydrochloride (1:2), 1-Chloro-3-(4-methyl-1-piperazinyl)propane Dihydrochloride, 1-Methyl-4-(3-chloropropyl)piperazine Dihydrochloride, 3-(4-Methylpiperazino)propyl Chloride Dihydrochloride, N-(3-Chloropropyl)-N'-methylpiperazine Dihydrochloride
InChI formel InChI=1S/C8H17ClN2.2ClH/c1-10-5-7-11(8-6-10)4-2-3-9;;/h2-8H2,1H3;2*1H
SMIL Cl.Cl.CN1CCN(CCCCl)CC1
IUPAC navn 1-(3-chloropropyl)-4-methylpiperazine;dihydrochloride
Molekylvægt (g/mol) 249.61