Få mere at vide
1-(3-Chloropropyl)-1,3-dihydrobenzimidazol-2-one (>85%), TRC
Artikelnummer. 30137218
Skift visning
Mængde:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137218 100 g
30012876 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137218 Leverandør Toronto Research Chemicals Leverandørnr. TRCC379825100G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 62780-89-6
Molekylær formel C10 H11 Cl N2 O
Molekylvægt (g/mol) 210.66
Synonym 2H-Benzimidazol-2-one, 1-(3-chloropropyl)-1,3-dihydro-, 1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one, 1-(3-Chloropropyl)-1,3-dihydrobenzimidazol-2-one
IUPAC navn 3-(3-chloropropyl)-1H-benzimidazol-2-one
SMIL ClCCCN1C(=O)Nc2ccccc12
Tekniske data
Kemisk navn eller materiale 1-(3-Chloropropyl)-1,3-dihydro-2H-benzimidazol-2-one
Mængde 100 g
InChI formel InChI=1S/C10H11ClN2O/c11-6-3-7-13-9-5-2-1-4-8(9)12-10(13)14/h1-2,4-5H,3,6-7H2,(H,12,14)
Formel vægt 210.056